Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-11
Entrez Gene ID
183610
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_C49D10.8
Chromosome Location
Chr.II: 3858142 - 3860926 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062273.1 NP_494674.1 9 - 10 1746 66

View Full Product Details