Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-40
Entrez Gene ID
190189
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y52E8A.5
Chromosome Location
Chr.II: 4314417 - 4316111 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377781.1 NP_001364687.1 3 - 4 459 124

View Full Product Details