Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-29
Entrez Gene ID
186488
Gene Name
Zinc metalloproteinase nas-29
Gene Aliases
CELE_F58A6.4
Chromosome Location
Chr.II: 5138519 - 5142036 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062552.5 NP_494953.3 7 - 8 1026 135

View Full Product Details