Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srb-16
Entrez Gene ID
186489
Gene Name
Serpentine receptor class beta-16
Gene Aliases
CELE_F58A6.6
Chromosome Location
Chr.II: 5148530 - 5150929 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062555.4 NP_494956.1 5 - 6 667 104

View Full Product Details