Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srb-1
Entrez Gene ID
191783
Gene Name
Serpentine receptor class beta-1
Gene Aliases
CELE_C27D6.10
Chromosome Location
Chr.II: 5169831 - 5171032 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062564.5 NP_494965.1 2 - 3 443 164

View Full Product Details