Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pmt-1
Entrez Gene ID
173901
Gene Name
protein-coding
Gene Aliases
CELE_ZK622.3
Chromosome Location
Chr.II: 5278090 - 5282525 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062589.9 NP_494990.2 1 - 2 49 66
NR_002630.1 1 - 2 41 66

View Full Product Details