Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vhp-1
Entrez Gene ID
173904
Gene Name
protein-coding
Gene Aliases
CELE_F08B1.1
Chromosome Location
Chr.II: 5323276 - 5340829 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356833.4 NP_001343753.1 1 - 1 197 152
NM_001377798.2 NP_001364748.1 1 - 2 188 152
GenBank mRNA AY585194.1 - 1 - 2 185 152

View Full Product Details