Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vhp-1
Entrez Gene ID
173904
Gene Name
protein-coding
Gene Aliases
CELE_F08B1.1
Chromosome Location
Chr.II: 5323276 - 5340829 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356833.4 NP_001343753.1 2 - 2 316 67
NM_001356834.3 NP_001343752.1 1 - 2 91 67
NM_001377798.2 NP_001364748.1 2 - 3 307 67
GenBank mRNA AY585194.1 - 2 - 3 304 67

View Full Product Details