Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vhp-1
Entrez Gene ID
173904
Gene Name
protein-coding
Gene Aliases
CELE_F08B1.1
Chromosome Location
Chr.II: 5323276 - 5340829 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356833.4 NP_001343753.1 7 - 7 1778 65
NM_001356834.3 NP_001343752.1 5 - 6 1553 65
NM_001356836.3 NP_001343755.1 2 - 3 1139 65
NM_001377798.2 NP_001364748.1 6 - 7 1769 65
NM_001377799.2 NP_001364848.1 3 - 4 1301 65
NM_062597.4 NP_494998.1 4 - 5 1823 65
GenBank mRNA AY585194.1 - 6 - 7 1766 65

View Full Product Details