Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ndx-6
- Entrez Gene ID
-
173916
- Gene Name
- Putative nudix hydrolase 6
- Gene Aliases
-
CELE_EEED8.8
- Chromosome Location
-
Chr.II: 5387455 - 5390104 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062614.5 |
NP_495015.1 |
7 - 8 |
1768 |
70 |
- Gene Symbol
- rsp-4
- Entrez Gene ID
-
173915
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_EEED8.7
- Chromosome Location
-
Chr.II: 5387411 - 5389884 on Build WBcel235
View Full Product Details