Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ifb-2
Entrez Gene ID
173973
Gene Name
protein-coding
Gene Aliases
CELE_F10C1.7
Chromosome Location
Chr.II: 5746731 - 5754143 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306596.3 NP_001293525.1 1 - 2 64 76
NM_062732.6 NP_495133.1 1 - 2 74 76
NM_062733.6 NP_495134.1 3 - 4 204 76

View Full Product Details