Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fzo-1
Entrez Gene ID
173990
Gene Name
Transmembrane GTPase fzo-1
Gene Aliases
CELE_ZK1248.14
Chromosome Location
Chr.II: 5833328 - 5836203 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062760.8 NP_495161.1 5 - 6 1635 87

View Full Product Details