Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nmgp-1
Entrez Gene ID
184437
Gene Name
Neuronal Membrane GlycoProtein
Gene Aliases
CELE_F13H8.4
Chromosome Location
Chr.II: 6260857 - 6262524 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062852.7 NP_495253.2 4 - 5 941 121

View Full Product Details