Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-10
Entrez Gene ID
183015
Gene Name
GLOBIN domain-containing protein
Gene Aliases
CELE_C29F5.7
Chromosome Location
Chr.II: 6308113 - 6310192 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062867.3 NP_495268.1 2 - 3 666 88

View Full Product Details