Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-31
Entrez Gene ID
183858
Gene Name
Serpentine receptor class alpha-31
Gene Aliases
CELE_C56C10.5
Chromosome Location
Chr.II: 6576974 - 6578478 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062931.3 NP_495332.2 4 - 5 848 187

View Full Product Details