Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ift-74
Entrez Gene ID
174102
Gene Name
protein-coding
Gene Aliases
CELE_C18H9.8
Chromosome Location
Chr.II: 6680024 - 6683544 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356863.3 NP_001343748.1 4 - 5 1042 94
NM_062958.6 NP_495359.2 5 - 6 1287 94

View Full Product Details