Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exos-8
Entrez Gene ID
174111
Gene Name
RNase_PH domain-containing protein
Gene Aliases
CELE_F41G3.14
Chromosome Location
Chr.II: 6741691 - 6742819 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062975.5 NP_495376.1 4 - 5 806 65

View Full Product Details