Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-44
Entrez Gene ID
174210
Gene Name
protein-coding
Gene Aliases
CELE_C07D10.2
Chromosome Location
Chr.II: 7394955 - 7397351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063148.6 NP_495549.1 2 - 3 158 66

View Full Product Details