Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cex-2
Entrez Gene ID
188309
Gene Name
protein-coding
Gene Aliases
CELE_T09A5.1
Chromosome Location
Chr.II: 7832838 - 7836615 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027157.5 NP_001022328.2 1 - 2 75 101
NM_001381511.2 NP_001366698.1 3 - 4 271 101
GenBank mRNA AY962814.1 - 1 - 2 75 101

View Full Product Details