Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubh-4
Entrez Gene ID
174289
Gene Name
Ubiquitin carboxyl-terminal hydrolase ubh-4
Gene Aliases
CELE_C08B11.7
Chromosome Location
Chr.II: 8038271 - 8040470 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383899.1 2 - 3 246 89

View Full Product Details