Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pch-2
Entrez Gene ID
174313
Gene Name
Putative pachytene checkpoint protein 2
Gene Aliases
CELE_F10B5.5
Chromosome Location
Chr.II: 8158349 - 8159925 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063310.9 NP_495711.1 3 - 4 771 77

View Full Product Details