Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nlp-5
Entrez Gene ID
185270
Gene Name
Neuropeptide-Like Protein
Gene Aliases
CELE_F35C11.1
Chromosome Location
Chr.II: 8238455 - 8243768 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063334.6 NP_495735.1 3 - 4 181 78

View Full Product Details