Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stc-1
Entrez Gene ID
174368
Gene Name
STCH (truncated HSP) family
Gene Aliases
CELE_F54C9.2
Chromosome Location
Chr.II: 8563709 - 8566139 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063407.6 NP_495808.2 2 - 3 180 130

View Full Product Details