Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atf-2
Entrez Gene ID
174399
Gene Name
ATF (cAMP-dependent transcription factor) family
Gene Aliases
CELE_K08F8.2
Chromosome Location
Chr.II: 8751986 - 8753940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063460.4 NP_495861.1 4 - 5 994 92

View Full Product Details