Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pah-1
Entrez Gene ID
174401
Gene Name
protein-coding
Gene Aliases
CELE_K08F8.4
Chromosome Location
Chr.II: 8762622 - 8766997 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267256.2 NP_001254185.1 1 - 2 178 80

View Full Product Details