Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wdfy-2
Entrez Gene ID
174475
Gene Name
WD repeat and FYVE domain-containing protein 2
Gene Aliases
CELE_D2013.2
Chromosome Location
Chr.II: 9319865 - 9321878 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063582.4 NP_495983.1 1 - 2 80 67

View Full Product Details