Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
die-1
Entrez Gene ID
174569
Gene Name
protein-coding
Gene Aliases
CELE_C18D1.1
Chromosome Location
Chr.II: 10061302 - 10067917 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063774.8 NP_496175.2 1 - 2 54 76

View Full Product Details