Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-2
Entrez Gene ID
191795
Gene Name
Serpentine receptor class delta-2
Gene Aliases
CELE_R05H5.1
Chromosome Location
Chr.II: 10185789 - 10187908 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063795.2 NP_496196.2 4 - 5 701 86

View Full Product Details