Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-59
Entrez Gene ID
191383
Gene Name
C-type LECtin
Gene Aliases
CELE_ZK666.5
Chromosome Location
Chr.II: 10478004 - 10479448 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063857.2 NP_496258.1 3 - 4 278 134

View Full Product Details