Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdl-1
Entrez Gene ID
174659
Gene Name
Histone RNA hairpin-binding protein
Gene Aliases
CELE_R06F6.1
Chromosome Location
Chr.II: 10785452 - 10787272 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063919.8 NP_496320.4 3 - 4 712 90

View Full Product Details