Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mix-1
Entrez Gene ID
174669
Gene Name
Mitotic chromosome and X-chromosome-associated protein mix-1
Gene Aliases
CELE_M106.1
Chromosome Location
Chr.II: 10824246 - 10841424 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063930.8 NP_496331.1 6 - 7 1643 72
GenBank mRNA U96387.1 - 6 - 7 1505 72

View Full Product Details