Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-5
Entrez Gene ID
187537
Gene Name
protein-coding
Gene Aliases
CELE_R03D7.6
Chromosome Location
Chr.II: 10950352 - 10951603 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267338.3 NP_001254267.1 1 - 2 199 62

View Full Product Details