Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-17
Entrez Gene ID
184514
Gene Name
Serpentine receptor class gamma-17
Gene Aliases
CELE_F15A4.7
Chromosome Location
Chr.II: 12473706 - 12475234 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064258.2 NP_496659.2 3 - 4 580 81

View Full Product Details