Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-62
Entrez Gene ID
174926
Gene Name
protein-coding
Gene Aliases
CELE_F35C5.5
Chromosome Location
Chr.II: 12893062 - 12895406 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064340.8 NP_496741.1 8 - 9 1186 77

View Full Product Details