Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lips-9
Entrez Gene ID
174935
Gene Name
LIPaSe related
Gene Aliases
CELE_F58G1.5
Chromosome Location
Chr.II: 12933854 - 12935362 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064355.4 NP_496756.1 3 - 4 493 70

View Full Product Details