Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-27
Entrez Gene ID
3564944
Gene Name
protein-coding
Gene Aliases
CELE_E01G4.4
Chromosome Location
Chr.II: 13448835 - 13460642 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383981.1 1 - 2 96 76

View Full Product Details