Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sre-45
Entrez Gene ID
186258
Gene Name
Serpentine receptor class epsilon-45
Gene Aliases
CELE_F54F11.3
Chromosome Location
Chr.II: 13517900 - 13522989 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064442.3 NP_496843.2 1 - 2 305 63

View Full Product Details