Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ing-3
Entrez Gene ID
3564936
Gene Name
Inhibitor of growth protein
Gene Aliases
CELE_Y51H1A.4
Chromosome Location
Chr.II: 13876034 - 13882585 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064508.5 NP_496909.1 3 - 4 232 53

View Full Product Details