Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gur-4
Entrez Gene ID
191664
Gene Name
GUstatory Receptor family
Gene Aliases
CELE_K09E4.5
Chromosome Location
Chr.II: 14104770 - 14108094 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064546.5 NP_496947.3 5 - 6 999 80

View Full Product Details