Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cacn-1
Entrez Gene ID
175066
Gene Name
protein-coding
Gene Aliases
CELE_W03H9.4
Chromosome Location
Chr.II: 14133320 - 14142168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267493.3 NP_001254422.1 4 - 5 939 79

View Full Product Details