Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hot-7
Entrez Gene ID
191682
Gene Name
Homolog of Odr-2 (Two)
Gene Aliases
CELE_Y48B6A.9
Chromosome Location
Chr.II: 14211418 - 14213315 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064563.3 NP_496964.2 1 - 2 116 77

View Full Product Details