Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plx-2
Entrez Gene ID
175101
Gene Name
Plexin-2
Gene Aliases
CELE_K04B12.1
Chromosome Location
Chr.II: 14417855 - 14424891 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377900.1 NP_001364609.1 11 - 12 4340 74

View Full Product Details