Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-28
Entrez Gene ID
190224
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_Y53F4B.31
Chromosome Location
Chr.II: 15166194 - 15167577 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064716.5 NP_497117.1 2 - 3 198 68

View Full Product Details