Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-1
Entrez Gene ID
190274
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y54F10BM.10
Chromosome Location
Chr.III: 2305926 - 2308126 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065133.3 NP_497534.1 2 - 3 835 140

View Full Product Details