Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-69
Entrez Gene ID
191826
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_H04J21.2
Chromosome Location
Chr.III: 2362948 - 2366278 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065141.1 NP_497542.1 3 - 4 429 91

View Full Product Details