Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbc-2
Entrez Gene ID
190267
Gene Name
RaBConnectin related
Gene Aliases
CELE_Y54F10AM.10
Chromosome Location
Chr.III: 2511640 - 2528689 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381780.2 NP_001368042.1 10 - 11 1693 78

View Full Product Details