Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pph-6
- Entrez Gene ID
-
183199
- Gene Name
- Serine/threonine-protein phosphatase 6 catalytic subunit
- Gene Aliases
-
CELE_C34C12.3
- Chromosome Location
-
Chr.III: 3461703 - 3467725 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065313.6 |
NP_497714.2 |
5 - 6 |
506 |
168 |
View Full Product Details