Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
toh-1
Entrez Gene ID
175491
Gene Name
Zinc metalloproteinase nas-26
Gene Aliases
CELE_T24A11.3
Chromosome Location
Chr.III: 3804772 - 3809030 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065368.6 NP_497769.3 4 - 5 805 104

View Full Product Details