Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnr-2
Entrez Gene ID
175525
Gene Name
Ribonucleoside-diphosphate reductase small chain
Gene Aliases
CELE_C03C10.3
Chromosome Location
Chr.III: 4089687 - 4094921 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065420.8 NP_497821.1 2 - 3 449 69

View Full Product Details