Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps-1
Entrez Gene ID
175584
Gene Name
40S ribosomal protein S3a
Gene Aliases
CELE_F56F3.5
Chromosome Location
Chr.III: 4475918 - 4476887 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065509.8 NP_497910.1 2 - 3 550 79

View Full Product Details